Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Allwik™ Emergency Spill Sak Portable Spill Kit, Universal

Catalog No. 17111278 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
10 gal./Bale
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-278 10 gal./Bale
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-278 Supplier Brady™ Supplier No. SKASAK
Add to Cart
Add to Cart

Allwik™ Emergency Spill Sak Portable Spill Kit, Universal provides a response for spills of oil, water-based and chemical liquids.

  • Absorbs up to 10 gallons per kit
  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids

Specifications

Color Black
Capacity (English) 10 gal./Bale
Material Polyethylene
Type Spill Sak Portable Spill Kit
For Use With (Application) Overspray leaks and drips, spill response
Includes 10 pads (15 x 19 in.), 2 SOCs (3 in. x 12 ft.), 1 pillow (17 x 19 in.), 1 disposal bag, 1 pair nitrile gloves, 1 instruction sheet
Dimensions (L x W x H) 32 x 20 x 4 in.
Width (Metric) 50.8 cm
Weight (English) 11.2 lb.
Weight (Metric) 24.7 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.