Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Allwik™ Large Spill Truck, Universal

Catalog No. 17111275 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
126 gal./Bale
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-275 126 gal./Bale
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-275 Supplier Brady™ Supplier No. SKALT
Add to Cart
Add to Cart

Allwik™ Large Spill Truck, Universal is designed for use with oil, water-based and chemical liquid spills.

  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
  • Large polyethylene bin is chemical and water resistant
  • Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills
  • Spill kits are ideal for fast response to critical accidents

Specifications

Color Yellow
Capacity (English) 126 gal./Bale
Material Polyethylene
Type Mobile Spill Kit
For Use With (Application) Overspray leaks and drips, spill response
Includes (200) 15 x 19 in. Pads, 3 in. x 4 ft. SOCs (24), 3 in. x 12 ft. SOCs (12), (8) 17 x 19 in. pillows, (3) pair nitrile gloves, (15) disposal bags, goggles, handbook
Dimensions (L x W x H) 47 x 34 x 43 in.
Width (Metric) 86.36 cm
Weight (English) 163.5 lb.
Weight (Metric) 360.52 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.