Learn More
Brady™ Allwik™ Portable Economy Spill Kit
Catalog No. 19000201 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W x H):
20 x 16 x 4 in.
Catalog No. Dimensions (L x W x H)
19-000-201 20 x 16 x 4 in.
1 options
Catalog No. 19-000-201 Supplier Brady™ Supplier No. SKAPP /DEL
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Includes
10 Pads (15 x 19 in.), 2 Socs (3 in. x 4 ft.), 1 Pair nitrile gloves, 1 Disposable bag, Instrution Sheet
Allwik™ Portable Economy Spill Kit provides a response for small spills of any material.
  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
  • Yellow PVC Bag offers high visibility spill control storage so items can be quickly located in an emergency situation
  • Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills
  • Portable spill kits are ideal for fast spill response
Specifications
Color Yellow
Type Allwik Portable Economy Spill Kit
Includes 10 pads, 2 SOCs, 1 pair nitrile gloves, 1 Disposable bag, Instrution Sheet
Dimensions (L x W x H) 20 x 16 x 4 in.
Width (Metric) 40.64 cm