Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Allwik™ RESCUE™ Kit, Universal

Catalog No. 17111210 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
20 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-210 20 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-210 Supplier Brady™ Supplier No. SKA20RESCUE

Allwik™ RESCUE™ Kit, Universal provides an effective spill response solution in an easy-to-use kit.

  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
  • 20-Gallon Drum is a UN-approved, water and chemical resistant polyethylene container with screw top lid for small-medium response
  • Kit includes “Caution” tape and a “Wet Floor” sign to help prevent slips, trips and falls during spill clean-up
  • Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills

Specifications

Color Yellow
Capacity (English) 20 gal.
Capacity (Metric) 75.7 L
Type Rescue Spill Control Kit, Universal
Includes 12 pads, 3 SOCs (3 x 12 in.), 2 pillows (18 x 18), 3 bags, 1 pair gloves, 1 pair goggles, 1 emergency response handbook, 1 roll of caution tape, 1 Wet Floor sign
Height (English) 18 in.
Diameter (English) 21 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.