Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Allwik™ Spill Kaddie Kit, Universal

Catalog No. 17111227 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
22 gal./Bale
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-227 22 gal./Bale
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-227 Supplier Brady™ Supplier No. SKAK2
Add to Cart
Add to Cart

Allwik™ Spill Kaddie Kit, Universal provides a fast response for spills, maintenance or mess prevention for any material.

  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
  • Mobile kaddie is water and chemical resistant and can be easily transported in case of emergency
  • Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills
  • Mobile kit can be deployed at a moment's notice when a spill occurs while the small footprint allows for storage in tight spaces

Specifications

Color Gray
Capacity (English) 22 gal./Bale
Material Polyethylene
Type Spill Kaddie Kit, Universal
For Use With (Application) Universal - All purpose spills
Includes Roll (1), SOCs (5), pillows (8), Pair Nitrile gloves (1), Disposal bags (5), goggles, emergency response handbook
Dimensions (L x W x H) 18 x 17 x 39 in.
Width (Metric) 43.18 cm
Weight (English) 41.5 lb.
Weight (Metric) 91.51 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.