Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Anti-Skid Hazard Marking Diecut Cleats

Catalog No. 19103787 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black, Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-103-787 Black, Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-787 Supplier Brady™ Supplier No. 78163
  • $248.00
    $168.65 / Pack of 24

    Save $79.35

    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use on steps and in entryways

  • Provides traction on slick floors
  • Printed message signals visual warning
  • Gives clear message to workers

Specifications

Color Black, Yellow
Width (Metric) 60.96 cm
Displayed Text Watch Your Step
Height (English) 6 in.
Height (Metric) 15.24 cm
Width (English) 24 in.
Material B-916 Grit-Coated Polyester
Type Anti-Skid Hazard Marking Diecut Cleats
Adhesive Type Permanent Pressure Sensitive Acrylic
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.