Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Anti-Slip Hazard Marking Tape

Catalog No. 19103824 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black, Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-103-824 Black, Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-824 Supplier Brady™ Supplier No. 92417
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used in marking dangerous areas of your plant

  • Contains printed message to caution employees about potential hazards

Specifications

Dimensions (L x W) 54 ft. x 3 in.
Color Black, Yellow
Finish Matte
Width (Metric) 7.62 cm
Width (English) 3 in.
Displayed Text Blank
Length (English) 54 ft.
Length (Metric) 16.4 m
Material B-819 Vinyl
Type Hazard Marking Tape
Adhesive Type Pressure Sensitive Acrylic
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.