Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Attack Pac - ReForm™ Spill Control Kit

Catalog No. 17111221 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
36 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-221 36 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-221 Supplier Brady™ Supplier No. SKRATK

ReForm™ Spill Control Kit Attack Pac is a space-saving, single-use kit packed in a puncture- and UV-resistant foil bag.

  • Attack Pac Portable Spill Kits combine absorbents, accessories, PPE and instruction guidelines so that you are prepared in an emergency
  • ReForm™ absorbents are made from 80% recycled newsprint and cellulose and are up to 50% more absorbent than comparable polypropylene absorbents
  • UV and puncture resistant bag includes a grommet for easy positioning in vehicles and manufacturing work cells
  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids

Specifications

Color Silver
Capacity (English) 36 gal.
Capacity (Metric) 136 L
Type Portable Spill Control Kit
Includes 15 pads (15 x 19 in), 5 SOCs (3 in. x 4 ft.), 1 pair of goggles, 1 pair of Nitrile gloves, (1) 6 mil plastic bag, 1 instruction sheet
Dimensions (L x W x H) 20 x 16 x 4 in.
Width (Metric) 40.64 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.