Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Battery Pack for Brady Mobile Printers

Catalog No. 19000631 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-000-631 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-000-631 Supplier Brady™ Supplier No. BMPUBP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Battery Pack for use with Brady™ BMP51 and BMP53 printers.

  • Package includes spare Li-Ion Printer Battery for the BMP51 and BMP53 printers only (label printer and AC Adaptor sold separately)
  • Long battery life - prints up to 1000 1 in. x 2 in. labels per battery charge
  • Use AC adapter/charger unit (not included) to charge battery
  • Enables cordless printer operation so you can print labels where you need them, when you need them

Specifications

Product Type Rechargeable Li-ION Battery
Color Gray
For Use With (Equipment) Brady Mobile Printers
Sterility Non-sterile
Alternate Part Number BMP-UBP 12V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.