Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BattleMat™ Tough Heavy Pads

Catalog No. 17111146 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
28 gal./Bale
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-146 28 gal./Bale
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-146 Supplier Brady™ Supplier No. BM100
Add to Cart
Add to Cart

BattleMat™ Tough Heavy Pads feature an industrial camouflage and a heavy duty approach to cleaning up spills.

  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
  • Heavy weight - specifically designed for continuous leaks and larger spills so you use less material
  • BattleMat™ absorbents are ideal as walk-on mats in manufacturing plants and similar environments
  • Perforations provide cost control and reduce waste by allowing you to use only what you need

Specifications

Color Gray
Capacity (English) 28 gal./Bale
Material Polypropylene
Type BattleMat Heavy Pad
For Use With (Application) Walkways, workstations, drums - liquid storage, outfalls and discharge Ponds
Packaging Quantity 100/Cs.
Weight Capacity Heavyweight
Dimensions (L x W) 38 x 48.26 cm (15 x 19 in.)
Length (Metric) 48.26 cm
Width (Metric) 38.1 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.