Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BBP™ Cutter Cleaning Tool

Catalog No. 19211173 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-211-173 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-211-173 Supplier Brady™ Supplier No. B31CCT
  • $32.50
    $22.65 / Each of 1

    Save $9.85

    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use the BBP™ Cutter Cleaning Tool to scrape residue off the cutting blade while cleaning your B30 Series Label Printer.

  • Package includes Cutter Cleaning Tool only (printer sold separately)
  • For use with B30 Series Label Printers, including the BBP™31 and BBP™33 Label Printers
  • Clean the printer cutter regularly to keep your label printer at optimal performance levels
  • Use a printer cleaning kit (not included) that includes Isopropyl Alcohol soaked soft foam swabs to wipe away any remaining residue

Specifications

Product Type Cutter Tool
Color Blue
For Use With (Equipment) BBP31 Sign and Label Printer
Sterility Non-sterile
Includes 1 Tool
For Use With (Application) Full ranges of labels for cable and wire ID, facility and safety ID, product ID and circuit board ID
Quantity 1 Ea.
Alternate Part Number B31-CCT
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.