Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BBP™ Dust Cover

Catalog No. 19211175 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-211-175 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-211-175 Supplier Brady™ Supplier No. B31DC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The optional BBP™ Dust Cover for the BBP™31 Sign and Label Maker protects your printer from environmental hazards when not in use.

  • Package includes printer cover only (printer sold separately)
  • Contoured design fits snuggly on the BBP™31 Sign and Label Maker
  • Protects your printer from dust and debris
  • Helps extend the life of your printer by keeping it clean and protected

Specifications

Product Type Dust Cover
Color Silver
For Use With (Equipment) BBP31 Sign and Label Printer
Dimensions (L x W x H) 6.25 x 6.25 x 6.25 in. (15.87 x 15.87 x 15.87 cm)
Sterility Non-sterile
Description BBP31 Dust Cover
Format Label
Quantity 1 Ea.
Alternate Part Number B31-DC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.