Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Biodegradable Flagging Tapes

Catalog No. 19104306 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black, Red
Black, Yellow
Displayed Text:
Caution
Danger
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Displayed Text
19-104-306 Black, Red Caution
19-104-304 Black, Yellow Danger
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-104-306 Supplier Brady™ Supplier No. 91090
  • $25.25
    $17.65 / Each of 1

    Save $7.60

    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used when hazardous marking is required in ecology conscious situations

  • Made of 70% wood fiber
  • Non-toxic
  • Will not harm wildlife, livestock or the environment
  • Complete degradation will occur in 6-12 months depending on environment conditions

Specifications

Color Black, Red
Length (English) 150 ft.
Length (Metric) 45.7 m
Width (Metric) 7.62 cm
Displayed Text Caution
Width (English) 3 in.
Type Biodegradable Flagging Tape
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.