Learn More
Brady™ BradyPrinter™ Model 3481
Catalog No. 19150178 Shop All Brady Corporation Products
Change view
Click to view available options
Bradyprinter 200m
Bradyprinter 200m/300
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-150-178 Bradyprinter 200m/300
19-150-177 Bradyprinter 200m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-150-178 Supplier Brady™ Supplier No. 56751
High-resolution 300 dpi graphics; Rugged enough for industrial settings; 12-month warranty on printer and printhead.
Rugged and reliable thermal transfer printer is ideal for desktop or industrial settings. Cost-effective and easy to use, with 300 dpi graphics, faster throughput (8 ips), and minimal label waste. Prints high-quality images on a variety of materials, from vinyl and polyester to polyimides. Image size ranges from as large as 4 in. (102mm) to as small as 1/8 in. (3.2mm). Frame is die-cast aluminum, with a modular design making it easy to change options, upgrade, or expand applications.
Warranty and Services
Printer and printhead come with a 12-month warranty.