Learn More
Brady™ BradyPrinter™ PR Plus Thermal Transfer Printer
Catalog No. 19211322 Shop All Brady Corporation Products
Change view
Click to view available options
Bradyprinter PR300 Plus Printer
Catalog No.
19-211-322 Bradyprinter PR300 Plus Printer
1 options
Catalog No. 19-211-322 Supplier Brady™ Supplier No. BPPR300+
BradyPrinter™ PR Plus Thermal Transfer Printer PR300+ is a high performance, 300 dpi THT printer for your cable ID needs and is ideal for printing Heatex products, wire markers, and polyester labels.
  • BradyPrinter™ PR300+ printer is perfect for use where material has a long life expectancy
  • Designed for up to 7000 labels per day at 12hrs / 5 days per week operation and prints at 300 dpi print resolution on labels up to 4" wide
  • Perfect for data communications labeling, general and industrial labeling, wire and cable marking, barcode labeling, and more
  • Includes the Brady360™ 5 year warranty that covers repair parts and labor, one print head replacement, a free loaner printer, a setup session, and more
Specifications
Product Type Bradyprinter™ PR300 Plus Printer
For Use With (Application) Barcode labeling, data communications labeling, general and industrial labeling, wire and cable marking
Dimensions (L x W x H) 10.8 x 9.5 x 17.6 in. (HxWxD)
Weight (English) 20 lb.