Learn More
Brady™ Bradystake™ Anchor
Catalog No. 19103880 Shop All Brady Corporation Products
Change view
Click to view available options
Packaging Quantity:
1 Ea.
Catalog No. Packaging Quantity
19-103-880 1 Ea.
1 options
Catalog No. 19-103-880 Supplier Brady™ Supplier No. 97084
  • $65.40
    $48.65 / Pack of 1
    Save $16.75
    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
For added stability in loose and sandy soil
  • Made of resilient, long-lasting spring steel
  • Catches soil around the stake preventing unauthorized removal
Order Info
As a custom order, anchors can be attached to Bradystake™ Warning Stakes at the factory for quick field installation
Specifications
Includes Anchors for five stakes, fasteners and instructions
Material Resilient spring steel
Type Anchor
Packaging Quantity 1 Ea.