Description
- Lightweight, portable pouch contains lockout devices to secure electrical equipment, including circuit breakers, electrical plugs and wall switches
- Convenient metal loops on the pouch allow pouch to hang on belt
- Equipping your employees with the proper lockout tools and warning devices can save lives, reduce lost employee time, and cut insurance costs
- Brady™ is a world leader in lockout/tagout products and services offering industry expertise and a broad range of durable, easy-to-use lockout devices
Specifications
Specifications
| Product Type | Breaker Lockout Pouch |
| Certifications/Compliance | OSHA 1910.147 |
| Depth (English) | 2 in. |
| For Use With (Equipment) | For electrical equipment |
| Includes | Pouch, small brady plug lockout, 3 wall switch lockouts, 3 universal multi-pole lockouts, 3 277V and 2 480/60Vv no-hole lockouts, 1 cleat 2 |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.