Learn More
Brady™ Breaker Lockout Sampler Pouch with Steel Padlocks and Tags
Shop All Brady Corporation ProductsDescription
- The Breaker Lockout Sampler Pouch contains one of each of our most popular circuit breaker lockouts
- Comes in a highly visible, easy to transport red pouch
- Equipping your employees with the proper lockout tools and warning devices can save lives, reduce lost employee time, and cut insurance costs
- Brady™ is a world leader in lockout/tagout products and services offering industry expertise and a broad range of durable, easy-to-use lockout devices
Specifications
Specifications
| Product Type | Breaker Lockout Sampler Pouch with Steel Padlocks and Tags |
| Certifications/Compliance | OSHA 1910.147 |
| Depth (English) | 2 in. |
| Includes | (1) 120V Snap-On Breaker Lockout, (1) 120/277V Clamp-On Breaker Lockout, (1) 480/600V Clamp-On Breaker Lockout, (1) Universal Multi-Pole Breaker Lockout, (1) Oversized Breaker Lockout, (1) Cleat for 120/277V Clamp-On Lockout, (1) Cleat for 480/600V Clamp-On Lockout, (1) Lockout Belt Pouch, 2-Brady Steel Padlocks and 2-Lockout Tags and nylon cable ties |
Safety and Handling
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.