Learn More
Brady™ Chemical Spill Control Absorbent Storage Cabinet Fill Kit
Catalog No. 17111001 Shop All Brady Corporation Products
Change view
Click to view available options
Chemical Spill Control Absorbent Storage Cabinet Fill Kit
Catalog No.
17-111-001 Chemical Spill Control Absorbent Storage Cabinet Fill Kit
1 options
Catalog No. 17-111-001 Supplier Brady™ Supplier No. SCCABINETHF
Absorbent Storage Cabinet is designed to provide storage and enable quick access to all of the essential absorbents for addressing chemical spills.
  • Spill Control Center Refill: Chemical, 26 gal
Specifications
Color Green
Type Chemical Spill Control Absorbent Storage Cabinet Refill Kit
Includes 15 x 19 in. pads (50), 3 in. x 4 ft. socks (4), 6 pillows, 5 disposal bags, 1 pair Nitrile gloves, goggles, handbook
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov