Learn More
Brady™ Chemical Spill Control Absorbent Storage Cart Fill Kit
Catalog No. 17111002 Shop All Brady Corporation Products
Change view
Click to view available options
Chemical Spill Control Absorbent Storage Cart Fill Kit
Catalog No.
17-111-002 Chemical Spill Control Absorbent Storage Cart Fill Kit
1 options
Catalog No. 17-111-002 Supplier Brady™ Supplier No. SCCARTHF
  • $607.00
    $450.65 / Each of 1
    Save $156.35
    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Absorbent Storage Cart is designed to provide storage and enable quick access to all of the essential absorbents for addressing chemical spills.
  • Spill Control Center Refill: Chemical, 32 gal
Specifications
Color Green
Type Chemical Spill Control Absorbent Storage Cart Refill Kit
Includes 15 x 19 in. pads (100), 3 in. x 4 ft. socks (8), 8 pillows, 5 disposal bags, 1 pair Nitrile gloves, goggles
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov