Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Connecting Hooks and Links

Catalog No. 19017252 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-017252 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-017252 Supplier Brady™ Supplier No. 78257
  • $160.00
    $78.65 / Pack of 36

    Save $81.35

    51% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Connecting S hooks and links from Bradylink.

  • The Bradylink Warning Post System is non-conductive so it can be safely used in electrically hazardous areas

Specifications

Color Yellow
For Use With (Application) Used to attaches lengths of warning chain
Width (English) 2 in.
Width (Metric) 5.08 cm
Material Polyethylene
Type S-Hook
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.