Brady™ Economy Pavement Marking Tape Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Brady™ 78261

 View more versions of this product

Catalog No. 19-046-628

  Check Availability
Add to Cart

    Specifications

    Format Tape
    For Use With (Application) Economical way to mark low traffic areas
    Length (English) 150 ft.
    Length (Metric) 45.7m
    Material Vinyl
    Adhesive Type Self-Adhesive
    Product Type Pavement Marking Tape
    Quantity Each
    Color Reflective White
    Width (English) 4 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit