Learn More
Description
Includes
Kit includes (pictured) 3/4 in. (1.9cm) blue padlocks (2), 2 in. (5.1cm) blue padlocks (2), lockout tags (10), red-labeled lockout hasps (3), small plug lockout (1), large plug lockout (1), single-pole breaker lockouts (6), 227V No Hole breaker lockouts (6), universal multipole breaker lockouts (6), wall switch lockouts (6).
Packed inside a bright yellow canvas carrying bag are the essential components of an electrical lockout/tagout safety system.
Order Info
Individual components available separately; contact your Fisher Safety Customer Service Representative.
Specifications
Specifications
| Product Type | Electrical Lockout Starter Kit |
| Color | Yellow |
| Material | Canvas |
| Certifications/Compliance | OSHA 1910.147 |
| Depth (English) | 2.5 in. |
| Depth (Metric) | 6 cm |
| For Use With (Application) | Protect employees from electrical hazards |
| Height (English) | 18 in. |
| Height (Metric) | 46 cm |
| Includes | Single pole breaker lockouts, 227 Volt No Hole breaker lockouts, universal multi pole breaker lockouts, wall switch lockouts, large Brady plug lockout, small Brady plug lockout, red labeled lockout hasps, 0.75 in. blue padlocks, 2 in. blue padlocks |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
