Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ ENV™ - MAXX Enhanced Oil Only Sorbent Pads

Catalog No. p-4933068 Shop All Brady Corporation Products
Click to view available options
Capacity (English):
24.5 gal./Bale
28 gal./Bale
33 gal./Bale
50 gal./Bale
51 gal./Bale
Weight Capacity:
Heavyweight
Lightweight
Medium-weight

Oil only sorbent equally effective on land or water

  • Single-ply construction
  • Repels water and floats indefinitely even when saturated
  • For use with oil and petroleum-based fluids
  • Available in two sizes
  • MAXX refers to enhanced performance without extra weight

    Applications:

    Marine applications, over spray leaks and drips, workstations, drums - liquid storage, outfalls and discharge ponds, emergency spill response.

    Specifications

    Color White
    Capacity (English) 24.5 gal./Bale
    Material Polypropylene
    Type Absorbent Pad, Oil-only
    For Use With (Application) Overspray leaks and drips, workstations, drums - liquid storage, outfalls and discharge ponds
    Packaging Quantity 100/Cs.
    Weight Capacity Lightweight
    Dimensions (L x W) 38.1 x 48.2 cm (15 x 19 in.)
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.