Learn More
Brady™ Extra Large Truck Spill Kit Refill, Chemical
Catalog No. 17111254 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
160 gal.
Catalog No. Capacity (English)
17-111-254 160 gal.
1 options
Catalog No. 17-111-254 Supplier Brady™ Supplier No. SKHXLTR
Extra-large mobile spill truck refill for hazardous chemical materials.
  • HAZWIK™ Mobile Spill Truck Refill Kit: Chemical, Extra Large
Specifications
Color Green
Capacity (English) 160 gal.
Capacity (Metric) 605.6 L
Type Chemical Spill Kit Refill, Extra-Large Truck
Includes 250 pads (15 x 19 in.), 36 SOCs (3 in. x 4 ft.), 20 SOCs (3 in. x 12 ft.), 8 pillows (18 x 18 in.), 30 disposal bags, 1 pair goggles, 3 pair nitrile gloves, emergency response guidebook