Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Flagging Tapes

Catalog No. p-6567152 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Blue
Fluorescent, Blue
Fluorescent, Orange
Fluorescent, Pink
Green
Orange
Red
White
Yellow
Length (English):
150 ft.
300 ft.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Length (English)
19-104-287 White 300 ft.
19-104-288 Green 300 ft.
19-104-289 Orange 300 ft.
19-104-290 Blue 300 ft.
19-104-291 Red 300 ft.
19-104-292 Yellow 300 ft.
19-104-295 Fluorescent, Blue 150 ft.
19-104-296 Fluorescent, Orange 150 ft.
19-104-298 Fluorescent, Pink 150 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-104-287 Supplier Brady™ Supplier No. 58342
Only null left
Add to Cart
Add to Cart

Used to mark boundaries and hazardous areas

  • Non-adhesive
  • Color-coded

Specifications

Dimensions (L x W) 300 ft. x 1.187 in.
Color White
Length (Metric) 91.4 m
Width (Metric) 3 cm
Type Flagging Tape
Length (English) 300 ft.
Width (English) 1.187 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.