Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Flip Style Vehicle General Commodity Placards, one-piece frame

Catalog No. 19110986 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
34.59 x 34.59cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-110-986 34.59 x 34.59cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-110-986 Supplier Brady™ Supplier No. 59955
  • $124.46
    $106.65 / Each of 1

    Save $17.81

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Legends:

Blank, Blasting Agents 1.5d, Corrosive 8, Dangerous, Dangerous When Wet 4, Explosive 1.4, Flammable 2, Flammable Gas 2, Flammable Solid 4, Inhalation 2, Inhalation 6 , Misc. 9, Non-Flammable Gas 2, Organic Peroxide 5.2, Oxidizer 5.1, Poison 6 , Spontaneously Combustible 4

For easy identification of chemical and hazardous materials. Flip Style, Vehicle General Commodity Placards, one-piece frame are approved for use by private and fleet carriers.

  • Self-contained placards slide easily on for aluminum hinges
  • Four stainless steel fasteners securely lock in chosen legend
  • Deep-formed aluminum frame provides stability and wind protection

Specifications

Material Aluminum
Quantity 1 Ea.
Height (English) 13.62 in.
Height (Metric) 34.59 mm
Width (English) 13.62 in.
Width (Metric) 34.59 cm
Shape Diamond
Dimensions (L x W) 34.59 x 34.59cm
Product Type Flip-style Placard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.