Learn More
Brady™ Flip Style Welding Supply and Medical Van Placards, one-piece frame
Shop All Brady Corporation ProductsDescription
Includes
Legends: Blank, Flammable Gas 2, Non-Flammable Gas 2 , Oxygen 2
- Standard flip-file system contain multiple panels in one frame
- Self-contained placards slide easily on for aluminum hinges
- Four stainless steel fasteners securely lock in your chosen legend
- Deep-formed aluminum frame provides stability and excellent wind protection
Specifications
Specifications
| Material | Aluminum |
| Quantity | 1 Ea. |
| Shape | Diamond |
| Languages | English |
| Product Type | Flip-style Placard |
| Color | Green, Red, Yellow |