Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Floor Safety Sign: CAUTION RADIATION AREA w/Picto

Catalog No. 19000420 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Vinyl
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-000-420 Vinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-000-420 Supplier Brady™ Supplier No. 47437

missing translation for 'validMainframeMsg'

Floor Safety Sign: CAUTION: RADIATION AREA with PICTOGRAM are designed to warn of hazardous areas.

  • Floor safety signs improve facility safety by alerting employees to hazards and/or important safety information in areas with foot traffic
  • B-819 Anti-Skid Vinyl features a velvet embossed rigid overlaminate
  • Excellent chemical, oil and water resistance
  • Minimum application temperature of 0°F (-18°C)

Specifications

Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
Material Vinyl
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Description Vinyl film with clear, matte, anti-slip overlaminate
For Use With (Application) Warn of hazardous areas, putting valuable safety information at the point of need
Thickness 0.0118 in.
Displayed Text Legend: CAUTION RADIATION AREA (W/Pictogram)
Product Type Floor Safety Sign
Color Pink on Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.