Learn More
Description
- Floor safety signs improve facility safety by alerting employees to hazards and/or important safety information in areas with foot traffic
- B-819 Anti-Skid Vinyl features a velvet embossed rigid overlaminate
- Excellent chemical, oil and water resistance
- Minimum application temperature of 0°F (-18°C)
Specifications
Specifications
| Certifications/Compliance | ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145 |
| Material | Vinyl |
| Quantity | 1 Ea. |
| Adhesive Type | Permanent Acrylic |
| Description | Vinyl film with clear, matte, anti-slip overlaminate |
| Diameter (English) | 17 in. |
| Diameter (Metric) | 43.1 cm |
| Thickness | 0.0118 in. |
| Languages | English |
| Displayed Text | EYE PROTECTION AREA (w/Pictogram) |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
