Learn More
MSC Brady Fuse Lockout, Fiberglass Nylon, 2 Padlocks, Red, 2 Pack
Catalog No. 5000677308
Encompass
This lockout device is designed for quick and easy installation with no tools needed. It positively locks out fuse blocks that previously could only be tagged. The device is made from durable and non-conductive fiberglass reinforced nylon. It accepts standard and compact safety padlocks and can also be locked out using a nylon tie.
  • Quick and easy installation with no tools needed.
  • Positively locks out fuse blocks that previously could only be tagged.
  • Made from durable and non-conductive fiberglass reinforced nylon.
  • Accepts standard and compact safety padlocks.
  • Can be locked out using a nylon tie.
Catalog No. 50-006-77308 Supplier MSC Supplier No. 10596344
This lockout device is designed for quick and easy installation with no tools needed. It positively locks out fuse blocks that previously could only be tagged. The device is made from durable and non-conductive fiberglass reinforced nylon. It accepts standard and compact safety padlocks and can also be locked out using a nylon tie.
  • Quick and easy installation with no tools needed.
  • Positively locks out fuse blocks that previously could only be tagged.
  • Made from durable and non-conductive fiberglass reinforced nylon.
  • Accepts standard and compact safety padlocks.
  • Can be locked out using a nylon tie.