Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ GHS Reference Center Poster

Catalog No. 19141908 Shop All Brady Corporation Products
Change view
Click to view available options
Width (Metric):
100.33 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
19-141-908 100.33 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-141-908 Supplier Brady™ Supplier No. 133209
  • $56.75
    $46.65 / Each of 1

    Save $10.10

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

All-inclusive poster combining important changes into one easy-to-read format

  • Large format
  • Details information on: new SDS format and sections, new GHS-formatted label elements and GHS Hazard Pictograms
  • Dimensions: 39.5W x 25.5 in. H

Specifications

Includes New safety data sheet (SDS) format and sections, new GHS-formatted label elements, GHS hazard pictograms
Material Paper
Quantity 1 Ea.
Mounting Type Wall Mount
For Use With (Application) Hazard communication regulation training
Height (English) 25.5 in.
Height (Metric) 64.77 cm
Width (English) 39.5 in.
Width (Metric) 100.33 cm
Languages English
Product Type GHS Reference Center Poster
Color White on Brown
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.