Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Globally Harmonized System (GHS) Flammable Picto Labels
GSA_VA

Catalog No. 19140513 Shop All Brady Corporation Products
Change view
Click to view available options
Quantity:
16/Pk.
39/Pk.
4/Pk.
Material:
Coated Paper
Vinyl
Height (English):
0.62 in.
1 in.
2 in.
Width (English):
0.6 in.
1 in.
2 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Material Height (English) Width (English)
19-140-513 39/Pk. Vinyl 0.62 in. 0.6 in.
19-140-522 39/Pk. Coated Paper 0.62 in. 0.6 in.
19-140-531 16/Pk. Vinyl 1 in. 1 in.
19-140-540 16/Pk. Coated Paper 1 in. 1 in.
19-140-549 4/Pk. Vinyl 2 in. 2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-140-513 Supplier Brady™ Supplier No. 118821
Only null left
Add to Cart
Edge
Add to Cart

For flammable gases, liquids and solids; pyrophorics; self-reatives; self-heating

Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards.

  • Legend: Flammable
  • Made of Vinyl or Coated paper
  • Available in three sizes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance GHS, OSHA 29 CFR 1910.1200
Adhesive Type Permanent Acrylic
Color Black, Red, White
Finish Gloss
Format Label
Material Vinyl
Product Type Pictogram Label
Sterility Non-sterile
Display Text Flammable
For Use With (Application) Flammable gases, liquids and solids
Quantity 39/Pk.
Temperature Range (English) -4°F to 167°F
Temperature Range (Metric) -20°C to +75°C
Dimensions (L x W) 0.625 x 0.625 in.
Width (English) 0.6 in.
Height (English) 0.62 in.
Height (Metric) 15.20 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.