Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ GlobalMark™ Label Printer: Stylus Pens

Catalog No. 19201247 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-201-247 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-201-247 Supplier Brady™ Supplier No. STYLUS3PK

Use these replacement stylus pens to select printer options easily and with precision on the touch screen of your GlobalMark™ Series Label Printer.

  • Package includes three replacement Stylus Pens (printer sold separately)
  • For use with the touch screen on GlobalMark™ Series Label Printers
  • This stylus pen flows smoothly over the surface of the screen
  • Navigation and selection are easy and accurate with a printer stylus

Specifications

Product Type Stylus Pen
Color Black
For Use With (Equipment) GlobalMark Printer
Sterility Non-sterile
Certifications/Compliance RoHS 2002/95/EC
Quantity 3/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.