Learn More
MSC Brady GuideStripe Tape, Yellow, 2 in, 100 ft, 1 Roll
Catalog No. 5000796803
Encompass
This GuideStripe Floor Tape is designed to identify indoor zones and safe distances, suitable for temporary applications due to its easy application and removal.
  • Ideal for areas with regular foot traffic
  • Vinyl material allows for easy application in straight lines
  • Strong acrylic adhesive suitable for marking areas within facilities
  • Resists chemicals, oil, and water
  • Can be applied with an applicator
  • Suitable for indoor use with service temperature range of -40 °F to 180 °F
Catalog No. 50-007-96803 Supplier MSC Supplier No. 13145792
This GuideStripe Floor Tape is designed to identify indoor zones and safe distances, suitable for temporary applications due to its easy application and removal.
  • Ideal for areas with regular foot traffic
  • Vinyl material allows for easy application in straight lines
  • Strong acrylic adhesive suitable for marking areas within facilities
  • Resists chemicals, oil, and water
  • Can be applied with an applicator
  • Suitable for indoor use with service temperature range of -40 °F to 180 °F