Description
- B-530 is ideal for use in asset identification, and applications where a deterrent to label removal is desired
- Breaks into small pieces during removal making it tedious to remove
- Service temperature range of -40 to 194° F
- Average outdoor durability of 3-5 years
Specifications
Specifications
| Color | White |
| Length (English) | 50 ft. |
| Width (English) | 0.5 in. |