Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazard Communication Handbooks

Catalog No. 19807197 Shop All Brady Corporation Products
Change view
Click to view available options
Product Type:
Hazard Communication Handbook
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-807-197 Hazard Communication Handbook
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-807-197 Supplier Brady™ Supplier No. 66206

Hazard Communication Handbooks help you train your employees in the identification of hazardous chemicals, use of product warning labels and understanding MSDS.

  • Handbooks also help you comply with OSHA's Hazard Communication Standard

Specifications

Format Book
Product Type Hazard Communication Handbook
For Use With (Application) Helps to identify the hazardous chemicals
Subject Safety and Training Materials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.