Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazardous Material Vehicle Placard

Catalog No. 19807141 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
White, Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-807-141 White, Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-807-141 Supplier Brady™ Supplier No. 63422
  • $72.30
    $56.65 / Each of 1

    Save $15.65

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hazardous Material Vehicle DOT Placards help ensure that hazardous material shipments reach their destinations without costly delays or rejections. All placards have square corners and no holes.

  • Placards meet D.O.T. requirements 49CFR Part 172
  • Use with slide-in Placard Holders from Brady™ (sold separately)
  • Graphics are permanently embedded within the sign panel, allowing for use in the most severe environments
  • Excellent chemical and abrasion resistance

Specifications

Certifications/Compliance ASTM D 1593, ASTM D 638, ASTM D 790, ASTM D 646
Adhesive Type Non-adhesive
Color White, Red
Finish Matte
Format Label
Material Fiberglass
Product Type Vehicle Placard
Sterility Non-sterile
For Use With (Application) Hazardous Material
Gloss 24 Gardner Units
Life Span 25 Years
Quantity 1 Ea.
Temperature Range (English) -40°F to 193°F
Temperature Range (Metric) -40°C to +90°C
Tensile Strength 9000 psi
Thickness (English) 0.1 in.
Dimensions (L x W) 10.75 x 10.75 in.
Width (English) 10.75 in.
Width (Metric) 27.305 cm
Height (English) 10.75 in.
Height (Metric) 27.3 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.