Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazardous Materials Labels

Catalog No. p-3521489 Shop All Brady Corporation Products
Change view
Click to view available options
Display Text:
Acetone
Solvent N.O.S.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
19-017668 Solvent N.O.S.
19-017659 Acetone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-017668 Supplier Brady™ Supplier No. 93735
Only null left
Add to Cart
Add to Cart

Hazardous Materials Labels are black-on-yellow vinyl labels that provide workers with all of the chemical information they will need for hazardous materials they may encounter.

  • The B-928 is a flexible vinyl film with a cold temperature applicable pressure sensitive adhesive
  • Service temperature range of -50 to 176°F
  • Average outdoor durability of 2 years
  • RoHS Compatibility Pass 2011/65/EU

Specifications

Certifications/Compliance RoHS
Adhesive Type Acrylic, Pressure-sensitive
Color Black, Yellow
Finish Matte
Format Label
Material Vinyl
Product Type Hazardous Waste Label
Sterility Non-sterile
Display Text Solvent N.O.S.
For Use With (Application) Hazardous waste material labels
Gloss 6 Gardner Units
Life Span 2 Years
Quantity 25/Pk.
Temperature Range (English) -50 to 176°F
Temperature Range (Metric) -46°C to 80°C
Tensile Strength 12 lb./in.
Thickness (English) 0.005 in.
Dimensions (L x W) 3.5 x 4.5 in.
Width (English) 4.5 in.
Width (Metric) 114.3 mm
Height (English) 3.5 in.
Height (Metric) 88.9 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.