Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazwik™ 20-Gal. Lab Pack Spill Response Kit

Catalog No. 19150269 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
17 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-150-269 17 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-150-269 Supplier Brady™ Supplier No. SKH20
  • $643.00
    $476.65 / Each of 1

    Save $166.35

    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UN-approved water- and chemical-resistant polyethylene containers with screw top

For small to medium response to hazardous materials.

Kit includes

  • (15) 38.1x 48.26cm (15 x 19 in.) pads
  • (3) 7.62Wcm x 3.66m (3 x 12 in.) SOCs
  • (2) 45.72 x 45.72cm (18 x 18 in.) pillows
  • Pair nitrile gloves, goggles and (3) disposal bags
  • Emergency response handbook
  • Sorbent Class: chemical
  • Absorbency: 16 gal./bale
  • Shipping wt.: 11.8kg (26 lbs.)

Compliance

UN approval

Specifications

Capacity (English) 17 gal.
Capacity (Metric) 64.26 L
Material Polyethylene
Type Drum Spill Kit
For Use With (Application) Small to medium response of a hazardous material
Includes (12) 15 x 19 in. pads, (3) 3 in. x 12 ft. socks, (2) 17 x 19 in. pillows, 1 pair nitrile gloves, three disposal bags, 1 pair goggles, emergency response handbook, screw top lid
Packaging Quantity 1 Kit
Length (English) 18 in.
Length (Metric) 45.72 cm
Diameter (English) 21 in.
Diameter (Metric) 53.34 cm
Weight (English) 26 lb.
Weight (Metric) 11.8 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.