Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazwik™ Chemical Absorbent Pillows

Catalog No. 19150274 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
18 x 18 in.
Packaging Quantity:
16/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Packaging Quantity
19-150-274 18 x 18 in. 16/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-150-274 Supplier Brady™ Supplier No. HAZ1818
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Surfactant-treated polypropylene designed for absorption of a wide range of chemicals, including hydrofluoric acid.

  • Chemically inert — will not react with aggressive fluids
  • Color-coded green indicates use with hazardous materials
  • Great for absorbing spills and leaking fluids in tight spaces
  • Class: chemical

HAZ1818

  • Shipping weight: 8.62kg (19 lbs.); 16 per case
  • HAZ99
    • Shipping weight: 5.44kg (12 lbs.); 32 per case

    Specifications

    Capacity (English) 28 gal.
    Capacity (Metric) 105.84 L
    Material Polypropylene
    Type Hazwik Sorbent Pillow
    For Use With (Application) Provide extra protection during packaging and transport, absorbs chemicals, acids and bases, spills and leaking fluids in tight spaces
    Packaging Quantity 16/Cs.
    Dimensions (L x W) 18 x 18 in.
    Weight (English) 19 lb.
    Weight (Metric) 8.6 kg
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.