Learn More
Brady™ Hazwik™ Emergency Spill Sak™ Portable Spill Kit, Chemical
Catalog No. 17111276 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
10 gal./Bale
Catalog No. Capacity (English)
17-111-276 10 gal./Bale
1 options
Catalog No. 17-111-276 Supplier Brady™ Supplier No. SKHSAK
Add to Cart
Add to Cart
Includes
10 pads (15 x 19 in.), 2 SOCs (3 in. x 12 ft.), 1 pillow (17 x 19 in.), 1 disposal bag, 1 pair nitrile gloves, 1 instruction sheet
For use with spills of hazardous materials
  • Absorbs up to 10 gal/bale
  • Portable economy spill kit packed in reusable sack
Specifications
Capacity (English) 10 gal./Bale
Material Polyethylene
Type Spill Sak Portable Spill Kit
Includes 10 pads (15 x 19 in.), 2 SOCs (3 in. x 12 ft.), 1 pillow (17 x 19 in.), 1 disposal bag, 1 pair nitrile gloves, instruction sheet
Weight (English) 9.5 lb.
Weight (Metric) 20.95 kg