Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazwik™ Spill Kaddie Kit, Chemical

Catalog No. 17111226 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
14 gal./Bale
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-226 14 gal./Bale
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-226 Supplier Brady™ Supplier No. SKHK2
Add to Cart
Add to Cart

Hazwik™ Spill Kaddie Kit, Chemical provides a fast spill response or maintenance mess prevention for hazardous chemical materials.

  • Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
  • Mobile kaddie is water and chemical resistant and can be easily transported in case of emergency
  • Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills
  • Mobile kit can be deployed at a moment's notice when a spill occurs while the small footprint allows for storage in tight spaces

Specifications

Color Gray
Capacity (English) 14 gal./Bale
Material Polyethylene
Type Spill Kaddie Kit, Chemical
For Use With (Application) Chemical spills
Includes Roll (1), SOCs (5), pillows (8), Pair Nitrile gloves (1), Disposal bags (5), goggles, emergency response handbook
Dimensions (L x W x H) 18 x 17 x 39 in.
Width (Metric) 43.18 cm
Weight (English) 41.5 lb.
Weight (Metric) 91.51 kg
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.