Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazwik™ Spill Kits

Catalog No. 19150272 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
5 gal.
15 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-150-272 15 gal.
19-150-268 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-150-272 Supplier Brady™ Supplier No. SKHCFB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical class spill kits

SKH-CFB Emergency Response Portable Spill Kit

  • Reusable clear bag with shoulder strap
  • (32) 38.1 x 48.26cm (15 x 19 in.) pads
  • (5) 7.62cm x 1.22m (3 in. x 4 ft.) SOCs
  • 43.18 x 48.26cm (17 x 19 in.) pillow
  • Nitrile gloves, goggles, two disposal bags and instruction sheet
  • Shipping wt: 11.34kg (25 lbs.)

SKH-PP — Portable Economy Spill Kit

  • For small spills of hazardous materials
  • (10) 38.1 x 48.26cm (15 x 19 in.) pads
  • (2) 7.62cm x 1.22m (3 in. x 4 ft.) SOCs
  • Nitrile gloves, disposal bag and instruction sheet
  • Shipping wt: 2.26kg (5 lbs.)

Specifications

Capacity (English) 15 gal.
Capacity (Metric) 56.7 L
Material Polyethylene
Type Portable Spill Containment Kit
For Use With (Application) Portable emergency response kit for overspray leaks and drips, spill response of hazardous materials
Includes (32) 38.1 x 48.26 cm pads, (5) 7.62 cm x 1.22 m socks, (1) 43.18 x 48.26 cm pillow, 1 pair nitrile gloves, goggles, 2 disposal bags, instructions
Packaging Quantity 1 Kit
Dimensions (L x W x H) 25.5 x 17 x 21.5 in. (64.77 x 43.18 x 54.61 cm)
Length (English) 25.5 in.
Length (Metric) 64.77 cm
Width (English) 17 in.
Width (Metric) 43.18 cm
Height (English) 21.5 in.
Height (Metric) 54.61 cm
Weight (English) 24.5 lb.
Weight (Metric) 54.0225 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.