Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazwik™ Spill Kits
GSA_VA

Catalog No. p-4492130 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
5 gal.
15 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-150-268 5 gal.
19-150-272 15 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-150-268 Supplier Brady™ Supplier No. SKHPP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical class spill kits

SKH-CFB Emergency Response Portable Spill Kit

  • Reusable clear bag with shoulder strap
  • (32) 38.1 x 48.26cm (15 x 19 in.) pads
  • (5) 7.62cm x 1.22m (3 in. x 4 ft.) SOCs
  • 43.18 x 48.26cm (17 x 19 in.) pillow
  • Nitrile gloves, goggles, two disposal bags and instruction sheet
  • Shipping wt: 11.34kg (25 lbs.)

SKH-PP — Portable Economy Spill Kit

  • For small spills of hazardous materials
  • (10) 38.1 x 48.26cm (15 x 19 in.) pads
  • (2) 7.62cm x 1.22m (3 in. x 4 ft.) SOCs
  • Nitrile gloves, disposal bag and instruction sheet
  • Shipping wt: 2.26kg (5 lbs.)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Color Yellow
Capacity (English) 5 gal.
Capacity (Metric) 15.12 L
Material PVC
Type Portable Spill Containment Kit
For Use With (Application) Portable economy kit for small spills
Includes (10) 38.1 x 48.26 cm pads, (2) 7.62 cm x 1.22 m socks, nitrile gloves, disposal bag, instruction sheet
Packaging Quantity 1 Kit
Dimensions (L x W x H) 20 x 16 x 4 in. (50.8 x 40.64 x 10.16 cm)
Length (English) 20 in.
Length (Metric) 50.8 cm
Width (English) 16 in.
Width (Metric) 40.64 cm
Height (English) 4 in.
Height (Metric) 10.16 cm
Weight (English) 3.909 lb.
Weight (Metric) 1.8 kg
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.