Learn More
Description
- Package includes the AC Adaptor (printer sold separately)
- For use with ID PAL™ and LabPal™ Series Label Printers
- Power supply formatted for North America
- With an AC power adapter on hand, you can be sure your printer is in optimal working condition when you need it the most
Specifications
Specifications
| Product Type | AC Adapter |
| Color | Black |
| For Use With (Equipment) | IDPAL Handheld Label Maker, LABPAL Handheld Label Maker |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.