Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ ID Safety Tapes

Catalog No. 19190003 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Magenta, Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-190-003 Magenta, Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-190-003 Supplier Brady™ Supplier No. 91464
Add to Cart
Add to Cart

ID Safety Tapes are durable, lightweight tapes made of non-adhesive, glossy B-912 polyethylene for on-the-spot warnings of potential danger.

  • Non-adhesive Polyethylene (B-912) is a glossy plastic that's ideal for temporary outdoor or permanent indoor use
  • Lightweight tape can be tied, stapled or nailed to posts, fences or metal barricades
  • Visual barricading provides fast on-the-spot warning of potential danger
  • B-912 polyethylene barricade tapes from Brady™ are available in variety of legends to fit your specific needs (each roll/legend is sold separately)

Specifications

Certifications/Compliance OSHA 1910.144
Adhesive Type Non-adhesive
Color Magenta, Yellow
Finish Gloss
Format Tape
Material Polyethylene
Product Type Tape
Sterility Non-sterile
Display Text Caution Radiation Hazard
For Use With (Application) For temporary outdoor or permanent indoor applications
Quantity 1 Ea.
Thickness (English) 0.003 in.
Dimensions (L x W) 1000 ft. x 3 in.
Length (English) 1000 ft.
Length (Metric) 304.8 m
Width (English) 3 in.
Width (Metric) 76.2 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.