Learn More
Brady™ Brady IP™ Printer - 600 DPI Standard with LabelMark™ Label Design Software
Catalog No. 19104489 Shop All Brady Corporation Products
Change view
Click to view available options
Brady IP™ Printer - 600 DPI Standard with LabelMark™ Label Design Software
Catalog No.
19-104-489 Brady IP™ Printer - 600 DPI Standard with LabelMark™ Label Design Software
1 options
Catalog No. 19-104-489 Supplier Brady™ Supplier No. BPIP600LMS
Brady IP™ Printer with LabelMark™ Standard Software redefines power and ease-of-use, making high performance, thermal transfer printing faster using smart chip technology.
  • Brady IP™ Printer's smart chip technology allows the printer, ribbon, material roll and PC software to communicate to reduce set up & change-out time
  • LabelMark™ Standard Software takes the hassle out of any labeling project from basic label design and printing to advanced import and editing options
  • No more calibration or material waste required for notched, gapped or continuous materials
  • Designed for up to 5000 labels per day at 12hrs / 5 days per week operation and prints up to 8 inches per second with 600 dpi print resolution
Specifications
Product Type Printer Kit
Autoclavable Not Autoclavable
Description Printer Kit
Includes BRADY IP™ 600 Printer
Quantity Kit