Learn More
Brady™ Jack Chain
Catalog No. 19113176 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
19-113-176 Stainless Steel
1 options
Catalog No. 19-113-176 Supplier Brady™ Supplier No. 98859
Add to Cart
Add to Cart
Made of stainless steel
  • Has open and close links to adjust to size needed
  • Contains approximately 25 links/ ft.
  • 10 to 12 in. is enough to fasten one tag
Specifications
Packaging Quantity 25 links/ft.
Length (English) 50 ft.
Material Stainless Steel
Length (Metric) 15.2 m
Type Jack chain