Learn More
Brady™ Keyed Alike Steel Padlocks with 2 inch Shackle Locks
Catalog No. 19103376 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Blue
Green
Red
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-103-376 Blue
19-103-378 Green
19-103-375 Red
19-103-381 Yellow
19-103-377 Yellow
19-103-380 Blue
19-103-382 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-103-376 Supplier Brady™ Supplier No. 105895
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes
2 keys
Hardened steel shackles withstand severe physical abuse and offer superior corrosion resistance
  • Made from reinforced laminated steel to withstand severe physical abuse and come with a hardened steel shackle making them extremely difficult to cut
  • Body plates are individually coated for superior rust protection
  • 5-pin cylinder offers more unique key cuts and better tamper resistance than conventional 4-pin locks
  • Lock body size is 1 1/3H x 1 9/16W x 7/8 in. D
  • Colored bumpers can be used for identification and protection a
  • Lock labels sold separately
  • When keyed alike, all locks in a set can be opened with the same key. This option is beneficial when multiple locks are assigned to a single employee
Specifications
Color Blue
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.